Likely not, due to pore size/possible fouling/chemical compatibility. Refer to our guide, "Chemical compatibility of filter components": https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/302/855/membrane-chemical-compatibility-ds2532en-ms.pdf
S2GPT01RE
Millipore® Steritop® Vacuum Bottle Top Filter
pore size 0.22 μm, polyethersulfone membrane, funnel capacity 150 mL, pack of 12 ea
Synonym(s):
SCGPT01RE, Steritop Threaded Bottle Top Filter, bottle top sterile filter unit, bottle top vacuum filter, bottle top vacuum filtering unit
About This Item
Recommended Products
material
polyethersulfone membrane
polystyrene funnel(s)
polystyrene housing
Quality Level
sterility
sterile; irradiated
product line
Steritop®
feature
hydrophilic
packaging
pack of 12 ea
parameter
45 °C max. temp.
description
2.4 mL (hold-up volume)
technique(s)
sterile filtration: suitable
H
74 mm
capacity
150 mL
diam.
12.5 cm
filter diam.
73 mm
filtration area
40 cm2
funnel capacity
150 mL
matrix
Express PLUS™
pore size
0.22 μm pore size
fitting
inlet funnel
45 mm outlet thread(s)
shipped in
ambient
General description
Stericup® and Steritop® sterile filtration devices use reliable, high-performance membranes for optimum sterile filtration of laboratory reagents, buffers, and media. These devices combine superior flow rates and throughput with low non-specific binding and a stable, no-tip design.
This Steritop® 150 mL bottle-top with 0.22 μm pore size polyethersulfone (PES) membrane filter unit can be used on bottles with 45 mm openings, designed for vacuum applications.
Application
- Tissue culture media +/- additives
- Buffers
- Biological solutions
Features and Benefits
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Evaluation of sterile filtration devices for protein preservation in sterile filtration of cell culture media.
Evaluation of sterile filtration devices for protein preservation in sterile filtration of cell culture media.
Evaluation of sterile filtration devices for protein preservation in sterile filtration of cell culture media.
Evaluation of sterile filtration devices for protein preservation in sterile filtration of cell culture media.
Related Content
This article shows that a PES membrane containing Stericup Quick Release filter provides an alternative for the pluripotent expansion of embryonic stem cells.
This article shows that a PES membrane containing Stericup Quick Release filter provides an alternative for the pluripotent expansion of embryonic stem cells.
This article shows that a PES membrane containing Stericup Quick Release filter provides an alternative for the pluripotent expansion of embryonic stem cells.
This article shows that a PES membrane containing Stericup Quick Release filter provides an alternative for the pluripotent expansion of embryonic stem cells.
-
Can we filter oil-type liquids with Stericup® filters?
Can we filter oil-type liquids with Stericup® filters?
1 answer-
Helpful?
-
-
What is the maximum processing volume for Stericup® and Steritop® filters? Can I add more volume after initial filtration?
1 answer-
All Stericup® and Steritop® filters were developed for single-use application. The maximum volume is dependent on the receiver bottle capacity. Stericup® filters come with a receiver bottle with a specified maximum volume. For Steritop® filters, the maximum recommended volume is 2 L with an appropriate-sized receiver bottle. Larger processing volumes can result in membrane fouling, increase chance of spillage and contamination, and lead to possible airlock*.
*Note that devices with PES membranes can airlock if filtered to dryness. Some liquid must be left on top of the filter, then the system stopped, receiver changed or emptied, and more sample volume added before restart. This increases the chance of spillage.Helpful?
-
-
Can Stericup® and Steritop® filters be reused?
1 answer-
All Stericup® and Steritop® filters were developed for single use, disposable processing.
Helpful?
-
-
Which type of pump should be used with Stericup® and Steritop® vacuum filters?
Which type of pump should be used with Stericup® and Steritop® vacuum filters?
1 answer-
We recommend our Chemical Duty Pump: https://www.sigmaaldrich.com/product/mm/wp6111560
Helpful?
-
-
Can Stericup® bottles be placed inside freezers for long-term storage of media?
1 answer-
You can successfully freeze and store many aqueous solutions (such as culture media) in Stericup® bottles at temperatures up to -20 °C (-4 °F). It is strongly recommended that you run a sample stability trial under your actual storage conditions prior to using Stericup® bottles for frozen storage.
Helpful?
-
-
Are Stericup® and Steritop® filters GMP-certified?
1 answer-
No, Stericup® and Steritop® devices are not GMP-approved.
Helpful?
-
-
The User Guide says “Use only glass or plastic bottles designed for vacuum applications”. Can you specify which bottles are compatible?
1 answer-
Steritop® Quick Release bottle top units require a vacuum-rated receiver bottle with either a GL33 or GL45 mm closure thread as specified. Steritop® E bottle top units require a vacuum-rated receiver bottle with a GL45 mm closure thread. Bottles no larger than 2 L should be used.
Glass bottles, GL33:
• Wheaton media bottles (glass), 500 mL (Product Z364886)
• Wheaton media bottles (glass), 1 L (Product Z364894)
Glass bottles, GL45
• Corning® glass media storage bottle, 125 mL (Product CLS1399125)
• Corning® glass media storage bottle, 250 mL (Product CLS1399250)
• Corning® glass media storage bottle, 500 mL (Product CLS1399500)
• Corning® glass media storage bottle, 1 L (Product CLS13991L)
• Pyrex® round media storage bottle, 500 mL (Product CLS1395500)
• Pyrex® round media storage bottle, 1 L (Product CLS13951L)
• KIMBLE® KIMAX® Media bottle, 250 mL (Product DWK14395250)
• KIMBLE® KIMAX® Media bottle, 500 mL (Product DWK14395500)
• KIMBLE® KIMAX® Media bottle, 1 L (Product DWK143951000)
Plastic (pre-sterile) bottles, GL45:
• Stericup® Quick Release Receiver flask, 250 mL (Product S200B02RE)
• Stericup® Quick Release Receiver flask, 500 mL (Product S200B05RE)
• Stericup® Quick Release Receiver flask, 1000 mL (Product S200B10RE)Helpful?
-
-
What leachables are detected during filtration processes using Stericup® filters?
1 answer-
We have not performed extractables/leachables studies for our Stericup® filters.
Helpful?
-
-
What Bacterial Endotoxins Test is used?
1 answer-
The test method used complies with USP 85 and EP 2.6.14 to ensure the absence of endotoxin in our products.
Helpful?
-
-
What is the difference between classic Stericup® filters and Stericup® “Quick Release” filters?
1 answer-
Stericup® Quick Release, or “QR”, filters feature a connection between funnel filter and receiver bottle that requires only a quarter-turn release to facilitate funnel removal. These filters come fully engaged – there is no need to tighten before use. The receiver bottle cap has flanges on the top surface to improve grip, as well as a positive lock stop to ensure a leak-proof seal with the receiver bottle. Other improvements include a large, frosted writing surface on the receiver bottle and a lighter cap color to facilitate labelling, more prominent volume markings on the filtration funnel and receiver bottle, and a product label with a peel-off lot record strip on the bag for easy viewing in storage.
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service