MWTC
mPAGE® Mini Wet Transfer Cassette
replacement part for mPAGE® Mini Wet Transfer System
All Photos(1)
About This Item
UNSPSC Code:
41105305
NACRES:
NA.32
Recommended Products
packaging
pkg of 1 pack
manufacturer/tradename
Millipore
technique(s)
SDS-PAGE: suitable
electrophoresis: suitable
compatibility
for use with mPAGE® Mini Wet Transfer System.
General description
The mPAGE® Mini Wet Transfer Cassette is a replacement part for the mPAGE® Mini Wet Transfer System (MWTS).
The mPAGE® Mini Wet Transfer System (MWTS) accommodates two gel holder cassettes for electrophoretic transfer of proteins from mini gels to PVDF or nitrocellulose blotting membrane using mPAGE® Transfer Buffer or a compatible transfer buffer.
The mPAGE® Mini Wet Transfer System (MWTS) accommodates two gel holder cassettes for electrophoretic transfer of proteins from mini gels to PVDF or nitrocellulose blotting membrane using mPAGE® Transfer Buffer or a compatible transfer buffer.
Legal Information
MPAGE is a registered trademark of Merck KGaA, Darmstadt, Germany