Skip to Content
Merck
All Photos(1)

Key Documents

MABT37

Sigma-Aldrich

Anti-Dentin sialoprotein Antibody, clone 2C12.3

clone 2C12.3, from mouse

Synonym(s):

Dentin sialophosphoprotein, Dentin phosphoprotein, Dentin phosphophoryn, DPP2.Dentin sialoprotein, DSP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2C12.3, monoclonal

species reactivity

mouse, rat

technique(s)

immunohistochemistry: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Dspp(666279)

General description

Dentin sialophosphoprotein (DSPP) is a protein of the extracellular matrix. It belongs to the small integrin-binding ligand, N-linked glycoprotein (SIBLING) family. DSPP is found in teeth and bone and is primarily involved in biomineralization processes in these structures. This function may be facilitated by the interaction of DSPP with multiple protein targets including DMP1, BMP2, Runx2, and NF-Y. DSPP plays a central role in dentinogenesis.

Immunogen

Pure DSP isolated rat dentin

Application

Anti-Dentin sialoprotein, clone 2C12.3 detects levels of Dentin sialoprotein proteins & has been published & validated for use in IHC.
Immunohistochemistry Analysis: A representative lot was used by an independent laboratory in rat mandibular tissue. (Baba, O., et al. (2004). Eur J Oral Sci. 112:163–170.)
Research Category
Cell Structure
Research Sub Category
Cytoskeleton

Quality

Evaluated by Immunohistochemistry in mouse and rat teeth tissues.

Immunohistochemistry Analysis: 1:250-1,000 dilution of this antibody detected Dentin sialophosphoprotein in mouse and rat teeth tissues.

Target description

68 kDa calculated. Uniprot describes 2 isoforms at 64 and 70 kDa

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2bκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Mouse and rat teeth tissues

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xin Gao et al.
Molecular medicine reports, 20(4), 3035-3042 (2019-08-23)
Dental epithelial‑mesenchymal signaling is crucial for tooth development, but the detailed mechanism is not fully understood. Using microarray analysis, it was revealed that the expression of osteoprotegerin, an important factor regulating bone remodeling, significantly increased after removal of the dental
Henry F Duncan et al.
Frontiers in cell and developmental biology, 10, 883266-883266 (2022-05-10)
Matrix-metalloproteinase-13 (MMP13) is important for bone formation and remodeling; however, its role in tooth development remains unknown. To investigate this, MMP13-knockout (Mmp13 -/- ) mice were used to analyze phenotypic changes in the dentin-pulp complex, mineralization-associated marker-expression, and mechanistic interactions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service