Skip to Content
Merck
All Photos(2)

Key Documents

MABS1824

Sigma-Aldrich

Anti-PTPRS Antibody, Clone 9H3.1

clone 9H3.1, from mouse

Synonym(s):

Receptor-type tyrosine-protein phosphatase S, EC: 3.1.3.48, R-PTP-S, Receptor-type tyrosine-protein phosphatase sigma, R-PTP-sigma

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

9H3.1, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... PTPRS(5802)

General description

Receptor-type tyrosine-protein phosphatase S (UniProt: Q13332; EC: 3.1.3.48, R-PTP-S, Receptor-type tyrosine-protein phosphatase sigma, R-PTP-sigma) is encoded by the PTPRS gene (Gene ID: 5802) in human. R-PTP-S is a single-pass type I membrane protein that belongs to the protein-tyrosine phosphatase family. Seven isoforms of R-PTP-S have been described that are produced by alternative splicing. R-PTP-S is detected in all tissues except plasma and liver. It is also detected in peripheral blood plasmacytoid dendritic cells. It is synthesized with a signal peptide (aa 1-29), which is subsequently cleaved off. The mature protein has an extracellular domain (aa 30-1282), a transmembrane domain (aa 1283-1303), and a cytoplasmic domain (aa 1304-1948). A cleavage is reported to occur between amino acids 1197-1198, which separates the extracellular domain from the transmembrane segment by a process known as ectodomain shedding. This process is involved in receptor desensitization, signal transduction and/or membrane localization. R-PTP-S serves as a cell surface receptor that binds to glycosaminoglycans, including chondroitin sulfate proteoglycans and heparan sulfate proteoglycan. Binding to chondroitin sulfate and heparan sulfate proteoglycans has opposite effects on R-PTP-S oligomerization and regulation of neurite outgrowth. It contributes to the inhibition of neurite and axonal outgrowth by chondroitin sulfate proteoglycans and plays a role in stimulating neurite outgrowth in response to the heparan sulfate proteoglycan GPC2. R-PTP-S is shown to be essential for normal brain development, especially for development of the pituitary gland and the olfactory bulb. It is reported to be rapidly internalized when dendritic cells are stimulated with the TLR9 ligand cytidine-phosphate-guanosine (CpG).

Specificity

Clone 9H3.1 detects Receptor-type tyrosine-protein phosphatase S (R-PTP-S) in human cells. It targets an epitope within 96 amino acids from the internal region.

Immunogen

GST/His-tagged recombinant fragment corresponding to 96 amino acids from the internal region of human Receptor-type tyrosine-protein phosphatase S (R-PTP-S). The immunogen sequence is conserved in all isoforms.

Application

Anti-PTPRS, Clone 9H3.1, Cat. No. MABS1824, is a mouse monoclonal antibody that detects Receptor-type tyrosine-protein phosphatase S and had been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.
Immunohistochemistry Analysis: A 1:1,000-2,000 dilution from a representative lot detected PTPRS in human tonsil, human lung, and human cervical cancer tissue sections.
Research Category
Signaling

Quality

Evaluated by Western Blotting in SH-SY5Y cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected PTPRS in 10 µg of SH-SY5Y cell lysate.

Target description

~220 kDa observed; 217.04 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service