Skip to Content
Merck
All Photos(2)

Key Documents

MABN356

Sigma-Aldrich

Anti-FKBP4 Antibody, clone 2H9.1

clone 2H9.1, from mouse

Synonym(s):

Peptidyl-prolyl cis-trans isomerase FKBP4, PPIase FKBP4, 51 kDa FK506-binding protein, FKBP51, 52 kDa FK506-binding protein, 52 kDa FKBP, FKBP-52, 59 kDa immunophilin, p59, FK506-binding protein 4, FKBP-4, FKBP59, HSP-binding immunophilin, HBI, Immunophi

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
2H9.1, monoclonal
application:
IHC
WB
species reactivity:
human
technique(s):
immunohistochemistry: suitable (paraffin)
western blot: suitable
citations:

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2H9.1, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... FKBP4(2288)

General description

FKBP4, also known as Peptidyl-prolyl cis-trans isomerase FKBP4 (PPIase FKBP4), or 51 kDa FK506-binding protein (FKBP51), 52 kDa FK506-binding protein (FKBP-52), or 59 kDa immunophilin (p59), or FK506-binding protein 4 (FKBP-4) or FKBP59 , or HSP-binding immunophilin (HBI), or Immunophilin FKBP52 Rotamase, and encoded by the gene FKBP4/FKBP62 is a chaperon type protein that is involved in protein folding and trafficking and the immunoregulation of cellular responses. FKBP4 is a member of the immunophilin family of proteins. FKBP4 forms a component of unligated steroid receptors complexes and helps in the cycling of the receptors between the cytoplasm and nuclear compartments. FKBP4 also plays a role in regulating neuronal growth cones via the regulations of TRPC1 channels, and FKBP4 modules microtubule dynamics by inhibiting MAPT/TAU in microtubule assembly. Finally, FKBP4 also influences the regulation and modulation oxidative stress in mitochondria. FKBP4 will localize to the cytoplasm, mitochondria or nucleus depending upon cellular situation and is widely expressed.

Immunogen

Epitope: C-terminus
GST-tagged recombinant protein corresponding to the C-terminus of human FKBP4.

Application

Anti-FKBP4 Antibody, clone 2H9.1 is an antibody against FKBP4 for use in western blotting & IHC (Paraffin).
Immunohistochemistry Analysis: A 1:500 dilution from a representative lot detected FKBP4 in human testis and human cerebellum tissue.
Research Category
Neuroscience
Research Sub Category
Developmental Signaling

Quality

Evaluated by Western Blotting in T47D cell lysate.

Western Blotting Analysis: 0.25 µg/mL of this antibody detected FKBP4 in 10 µg of T47D cell lysate.

Target description

~52 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service