Skip to Content
Merck
All Photos(1)

Key Documents

MAB3306

Sigma-Aldrich

Anti-MMP-3 Antibody, cross reacts w/hMMP-10, clone 55-2A4

clone 55-2A4, Chemicon®, from mouse

Synonym(s):

Stromelysin-1, Transin, Proteoglycanase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

55-2A4, monoclonal

species reactivity

human, pig, rat, sheep

should not react with

bovine, rabbit, mouse

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... MMP3(4314)

Specificity

The antibody specifically reacts with precursor and active forms of human MMP-3. This is a purified mouse monoclonal antibody to human matrix metalloproteinase 3 (human MMP-3, stromelysin-1) from synovial cells. The antibody cross reacts 34% with human MMP-10.

Immunogen

Epitope: cross reacts w/hMMP-10
Human matrix metalloproteinase 3 (human MMP-3, stromelysin-1) from synovial cells.

Application

Immunoblotting (0.5 μg/mL)

Immunohistochemistry on paraffin-embedded tissues (5-10μg/mL, PLP fixation required, not recommended for traditional formalin fixation. Immunohistochemistry on fresh frozen sections (0.5 μg/mL)

ElA
Research Category
Cell Structure
Research Sub Category
MMPs & TIMPs
This Anti-MMP-3 Antibody, cross reacts w/hMMP-10, clone 55-2A4 is validated for use in ELISA, WB, IH(P) for the detection of MMP-3.

Physical form

Format: Purified
Liquid in 0.01M Na-Phosphate buffer, pH 7.0, 125 mM NaCl, containing 2% protease free bovine serum albumin.

Storage and Stability

Maintain frozen at -20°C in undiluted aliquots for up to 12 months.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Manufactured by Daiichi Fine Chemical Co., Ltd

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Iraklis Papageorgiou et al.
Nanomaterials (Basel, Switzerland), 4(2), 485-504 (2014-06-16)
Medical interventions for the treatment of spinal disc degeneration include total disc replacement and fusion devices. There are, however, concerns regarding the generation of wear particles by these devices, the majority of which are in the nanometre sized range with
M Fréchet et al.
Oncogene, 27(39), 5223-5232 (2008-05-13)
Xeroderma pigmentosum (XP) is a rare, recessively inherited genetic disease characterized by skin cancer proneness and premature aging in photoexposed area. The disease results from defective nucleotide excision repair of ultraviolet (UV)-induced DNA lesions. Reconstruction of group C (XP-C) skin
Kalu U E Ogbureke et al.
Kidney international, 68(1), 155-166 (2005-06-16)
Three members of the small integrin-binding ligand N-linked glycoprotein (SIBLING) family of proteins have recently been shown to bind and activate specific promatrix metalloproteinases (MMPs) and to overcome the inhibition of tissue inhibitors of MMPs (TIMPs). Although usually associated with
Tracy N Clevenger et al.
Journal of tissue engineering, 7, 2041731416670482-2041731416670482 (2016-10-14)
One of the most common regenerative therapies is autologous fat grafting, which frequently suffers from unexpected volume loss. One approach is to deliver adipose stem cells encapsulated in the engineered hydrogels supportive of cell survival, differentiation, and integration after transplant.
Allison L Arai et al.
Biochemistry, 59(32), 2922-2933 (2020-07-24)
Matrix metalloprotease (MMP) activation contributes to the degradation of the extracellular matrix (ECM), resulting in a multitude of pathologies. Low-density lipoprotein receptor-related protein 1 (LRP1) is a multifaceted endocytic and signaling receptor that is responsible for internalization and lysosomal degradation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service