Skip to Content
Merck
All Photos(1)

Key Documents

EZMADP

Millipore

Mouse Adiponectin ELISA Kit

measures and quantifies Adiponectin levels in 20 μL serum, plasma or cell culture supernatent

Synonym(s):

Interferon beta-2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.32

Product Name

Mouse Adiponectin ELISA, This Mouse Adiponectin ELISA is used to measure & quantify Adiponectin levels in Metabolism & Endocrine research.

Quality Level

species reactivity

mouse

packaging

kit of 1 × 96 wells

parameter

20 μL sample volume (4hr assay)

assay range

accuracy: 99-112%
linearity: 96-121%
sensitivity: 0.78 ng/mL
standard curve range: 0.78-50 ng/mL

technique(s)

ELISA: suitable

input

sample type plasma (K2 EDTA)
sample type serum
sample type cell culture supernatant

UniProt accession no.

application(s)

research use

detection method

colorimetric (450nm/590nm)

shipped in

wet ice

storage temp.

2-8°C

Gene Information

mouse ... ADIPOQ(11450)

General description

Adiponectin is identified as a 30 kDa adipokine that is secreted from the adipose tissue into circulating blood and influences systemic metabolism.

Application

Research Category
Metabolism
Research Sub Category
Obesity

Metabolic Disorders
Room temperature, 3 hour assay 20 μL sample size (serum/plasma: 1:1000 dilution)
Used to detect/quantify: Adiponectin

Linkage

Replaces: CYT286

Disclaimer

For research use only. Not for use in diagnostic procedures.

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Met. Corr. 1 - Skin Sens. 1 - STOT RE 2 Oral

Target Organs

Brain

Supplementary Hazards

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Green tea extract protects leptin-deficient, spontaneously obese mice from hepatic steatosis and injury.
Richard S Bruno, Christine E Dugan, Joan A Smyth, Dana A DiNatale, Sung I Koo
The Journal of Nutrition null
The starvation hormone, fibroblast growth factor-21, extends lifespan in mice.
Zhang, Yuan, et al.
eLife, 1, e00065-e00065 (2012)
Timothy M Griffin et al.
Arthritis and rheumatism, 64(2), 443-453 (2011-09-29)
To test the hypotheses that obesity due to a very high-fat diet induces knee osteoarthritis (OA), and that short-term wheel-running exercise protects against obesity-induced knee OA by reducing systemic inflammation and metabolic dysregulation. Male C57BL/6J mice were fed either a
Swimming training beneficial effects in a mice model of nonalcoholic fatty liver disease.
Schultz A, Mendonca LS, Aguila MB, Mandarim-de-Lacerda CA
Experimental & Toxicologic Pathology null
Antidiabetic activity of resveratrol, a known SIRT1 activator in a genetic model for type-2 diabetes.
Sharma S, Misra CS, Arumugam S, Roy S, Shah V, Davis JA, Shirumalla RK, Ray A
Phytotherapy Research null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service