Skip to Content
Merck
All Photos(1)

Key Documents

AB5894

Sigma-Aldrich

Anti-P2X2 Receptor Antibody

serum, Chemicon®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

guinea pig

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat, human

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... P2RX2(22953)

Specificity

P2X2 Receptor.

Immunogen

A 13 amino acid peptide corresponding to amino acids 460-472 from the carboxy-terminus of the rat P2X2 Receptor protein.

Control peptide: Catalog Number AG354

Application

Anti-P2X2 Receptor Antibody is an antibody against P2X2 Receptor for use in IC, IH & WB.
Immunoblotting: 1:500

Immunohistochemistry: 1:500.

Immunocytochemistry: 1:500.

Optimal working dilutions must be determined by end user.

APPLICATION NOTES FOR AB5894

IMMUNOHISTOCHEMISTRY

Male Sprague-Dawley rats (b.wt. 100-150g) were anesthetized with sodium pentobarbital and perfused via the ascending aorta with: 1) 50 mL of Ca2+-free Tyrode+s solution followed by 2) a formalin-picric acid fixative (4% paraformaldehyde with 0.4% picric acid in 0.16 M phosphate buffer, pH 6.9) and 3) 10% sucrose in PBS as a cryo-protectant. Tissues were rapidly dissected out and stored overnight in 0.1 M phosphate buffer (pH 7.4) containing 10% sucrose. Slide-mounted tissue sections were incubated with blocking buffer for 1 hour at room temperature. Primary antibody was diluted in blocking buffer to the appropriate working dilution. Blocking buffer was removed and the slides were then incubated at 2-8°C for 18-24 hours with AB5894 (1:500). After rinsing in PBS 3 times sections were incubated for 60 minutes at room temperature with Cy3-conjugated secondary antibodies. After mounting in a mixture of PBS and glycerol (1:3) containing 0.1% p-phenylenediamine, sections were examined with a Nikon Microphot-SA epifluorescence microscope.

IMMUNOCYTOCHEMISTRYP

2X2 transfected cells were processed for indirect immunofluorescence. Media was removed and cells were gently washed 3 times with serum-free media. Following fixation procedure, cells were processed for indirect immunofluorescence as above

WESTERN BLOTTING

Cell membrane extracts were examined by electrophoresis (8% acrylamide) with SDS under reducing conditions and transferred to a nylon membrane. Membranes were blocked for 1 hour at 2-8°C with 0.1% Tween 20 and 2.5% milk powder (w/v) in PBS. Membranes were incubated with AB5894 diluted 1:500 with same buffer overnight at 2-8°C. Membranes were rinsed and incubated with HRP conjugated secondary antibody for 1 hour at room temperature. Following rinsing the membranes were processed using enhanced chemiluminescence.
Research Category
Neuroscience
Research Sub Category
Neurotransmitters & Receptors

Physical form

Serum. Liquid. Contains 0.05% sodium azide.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S E Mondello et al.
Experimental neurology, 335, 113480-113480 (2020-09-30)
To date, relatively few studies have used optogenetic stimulation to address basic science and therapeutic questions within the spinal cord. Even less have reported optogenetic stimulation in the rat spinal cord. This is likely due to a lack of accessible
Z-J Wang et al.
Anatomy and embryology, 207(4-5), 363-371 (2003-11-19)
Intraganglionic laminar endings (IGLEs) represent the most prominent vagal afferent terminal structures throughout the gastrointestinal tract. They are most prominent in the esophagus and stomach, but can be found down to the distal colon. Their role as mechanosensors as proposed
Drew L Sellers et al.
Proceedings of the National Academy of Sciences of the United States of America, 113(9), 2514-2519 (2016-02-19)
A significant unmet need in treating neurodegenerative disease is effective methods for delivery of biologic drugs, such as peptides, proteins, or nucleic acids into the central nervous system (CNS). To date, there are no operative technologies for the delivery of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service