Skip to Content
Merck
All Photos(3)

Key Documents

AB2984

Sigma-Aldrich

Anti-Integrin beta 3 (ITGB3/CD61) Antibody

rabbit polyclonal

Synonym(s):

integrin, beta 3 (platelet glycoprotein IIIa, antigen CD61), Platelet membrane glycoprotein IIIa, integrin beta-3, platelet glycoprotein IIIa, CD61 antigen

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

Product Name

Anti-Integrin beta-3 Antibody, from rabbit, purified by affinity chromatography

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, pig

species reactivity (predicted by homology)

rat (based on 100% sequence homology), canine (based on 100% sequence homology), porcine (based on 100% sequence homology), chimpanzee (based on 100% sequence homology), bovine (based on 100% sequence homology)

technique(s)

immunohistochemistry: suitable (paraffin)
immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ITGB3(3690)

General description

Integrin beta-3 is a member of the Integrin receptor family. The integrin family has been shown to be instrumental in cell adhesion to extracellular matrix proteins and can be found in certain cell to cell adhesions. Integrins are also important in transmembrane connections and intracellular signaling. Integrins are key to understanding many human diseases as they are commonly the receptors. Integrins have been linked to many different parts of cellular operations including; development, immunity, leukocyte traffic, homeostasis and even cancer. Once integrin beta-3 is activated, it binds fibrinogen which causes platelet to platelet interaction effectively cutting off a ruptured endothelial surface.

Specificity

Other homologies: Mouse (93% sequence homology.)
This antibody recognizes the extracellular domain of integrin beta-3.

Immunogen

Epitope: Extracellular Domain
KLH-conjugated linear peptide corresponding to human integrin beta-3 at the extracellular domain.

Application

Anti-Integrin beta-3 Antibody is an antibody against Integrin beta-3 for use in WB, IP, IH(P).
Immunohistochemistry Analysis: A 1:500 dilution from a previous lot detected intergin beta-3 in skin membrane tissue.

Immunoprecipitaiton Analysis: 10 µg from a previous lot immunoprecipitated integrin beta-3 from U251 RIPA lysate.
Research Category
Cell Structure
Research Sub Category
ECM Proteins

Quality

Evaluated by Western Blot in U251 cell lysate.

Western Blot Analysis: 0.5 µg/mL of this antibody detected integrin beta-3 in 10 µg of U251 cell lysate.

Target description

~85 kDa observed.
Uncharacterized bands may be observed in some lysates.

Linkage

Replaces: 04-1060

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
U251 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Naohiro Miyashita et al.
HemaSphere, 3(4), e268-e268 (2019-11-15)
We retrospectively evaluated 48 essential thrombocythemia (ET) patients who were treated in our institute (male/female, 14/34, median age, 61.5 years). In 14 patients treated with anagrelide (ANA), the degree of platelet count reduction (median, -56.6%) was strongly correlated with increase
Naohiro Miyashita et al.
Annals of hematology, 97(4), 629-640 (2018-01-31)
We identified a novel heterozygous ITGB3 p.T720del mutation in a pedigree with macrothrombocytopenia exhibiting aggregation dysfunction. Platelet aggregation induced by ADP and collagen was significantly reduced, while ristocetin aggregation was normal. Integrin αIIbβ3 was partially activated in a resting status
Ayuko Hoshino et al.
Nature, 527(7578), 329-335 (2015-11-03)
Ever since Stephen Paget's 1889 hypothesis, metastatic organotropism has remained one of cancer's greatest mysteries. Here we demonstrate that exosomes from mouse and human lung-, liver- and brain-tropic tumour cells fuse preferentially with resident cells at their predicted destination, namely
Ravi Chakra Turaga et al.
Nature communications, 7, 11675-11675 (2016-06-01)
Integrin αvβ3 expression is altered in various diseases and has been proposed as a drug target. Here we use a rational design approach to develop a therapeutic protein, which we call ProAgio, that binds to integrin αvβ3 outside the classical
D F Young et al.
The Journal of general virology, 90(Pt 11), 2731-2738 (2009-07-25)
Although the Enders strain of mumps virus (MuV) encodes a functional V protein that acts as an interferon (IFN) antagonist, in multi-cycle growth assays MuV Enders grew poorly in naïve ('IFN-competent' Hep2) cells but grew to high titres in 'IFN-compromised'

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service