Skip to Content
Merck
All Photos(1)

Key Documents

8.55094

Sigma-Aldrich

Trichloroacetimidate Wang resin

Novabiochem®

Synonym(s):

Trichloroacetimidate Wang resin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.22
form:
beads

Quality Level

product line

Novabiochem®

form

beads

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

storage temp.

2-8°C

SMILES string

ClC(Cl)(Cl)C(=N)OCc1ccc(cc1)OC

InChI

1S/C10H10Cl3NO2/c1-15-8-4-2-7(3-5-8)6-16-9(14)10(11,12)13/h2-5,14H,6H2,1H3

InChI key

TYHGKLBJBHACOI-UHFFFAOYSA-N

General description

A highly versatile reagent for the solid phase immobilization of alcohols [1,2,3]; carboxylic, phosphonic and sulfonic acids [4]; thiols and thioacids [4]; and phenols [5]. In contrast to the analogous bromide resin which is loaded under basic conditions, derivatization of this support is achieved under acidic conditions, making it compatible with base-sensitive compounds such as Fmoc-amino alcohols.

Associated Protocols and Technical Articles
Protocols for Loading of Peptide Synthesis Resins

Literature references

[1] S. Hanessian & X. Fang (1998) Tetrahedron Lett., 39, 733.
[2] A. Bianco, et al. (2000) J. Comb. Chem., 2, 681.
[3] A. Kamal, et al. (2002) Tetrahedron Lett., 43, 2103.
[4] C. W. Phoon, et al. (1998) Tetrahedron Lett., 39, 7959.
[5] A. Kamal, et al. (2001) Tetrahedron Lett., 42, 6969.

Linkage

Replaces: 01-64-0314

Analysis Note

Color (visual): slight yellow to beige
Appearance of substance (visual): beads
Loading (determined by elemental analysis after derivatization with thioacetic acid): 0.60 - 1.20 mmol/g
Swelling Volume (in DMF): lot specific result
The polymer matrix is copoly (styrene-1% DVB), 100 - 200 mesh

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service