Skip to Content
Merck
All Photos(5)

Key Documents

8.22275

Supelco

N,N-Dimethylformamide

EMPLURA®

Synonym(s):

DMF, N,N-Dimethylmethanamide, Formic acid dimethylamide, DMF, NSC 5356

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCON(CH3)2
CAS Number:
Molecular Weight:
73.09
Beilstein:
605365
MDL number:
UNSPSC Code:
12352111
EC Index Number:
200-679-5
NACRES:
NA.05
bp:
153 °C (lit.)
vapor pressure:
2.7 mmHg ( 20 °C)
Pricing and availability is not currently available.

vapor density

2.5 (vs air)

Quality Level

vapor pressure

2.7 mmHg ( 20 °C)

product line

EMPLURA®

Assay

≥99% (GC)

form

liquid

autoignition temp.

833 °F

potency

3010 mg/kg LD50, oral (Rat)
1500 mg/kg LD50, skin (Rabbit)

expl. lim.

15.2 %

dilution

(low-cost alternative, for basic applications)

impurities

≤0.1% Water

refractive index

n20/D 1.430 (lit.)

pH

7 (20 °C, 200 g/L in H2O)

bp

153 °C (lit.)

mp

−61 °C (lit.)

transition temp

flash point 57.5 °C

density

0.944 g/mL (lit.)

storage temp.

2-30°C

SMILES string

[H]C(=O)N(C)C

InChI

1S/C3H7NO/c1-4(2)3-5/h3H,1-2H3

InChI key

ZMXDDKWLCZADIW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

EMPLURA® is our low-cost alternative to high-purity qualities. With EMPLURA we offer a range of solvents for a plurality of basic applications in non-regulated industries and for less demanding applications, preparative laboratory work and cleaning purposes. EMPLURA solvents provide adequate specifications with the most important parameters.

Application

Solvent for many hydrophobic organic compounds.

Analysis Note

Purity (GC): ≥ 99.0 %
Identity (IR): conforms
Density (d 20 °C/ 4 °C): 0.948 - 0.949
Water: ≤ 0.10 %

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Liq. 3 - Repr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

135.5 °F - closed cup

Flash Point(C)

57.5 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service