Skip to Content
Merck
All Photos(1)

Key Documents

8.21902

Sigma-Aldrich

3-Methylbenzoic acid

for synthesis

Synonym(s):

3-Methylbenzoic acid, m-Toluic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
3-(CH3)C6H4COOH
CAS Number:
Molecular Weight:
136.15
MDL number:
UNSPSC Code:
12352106
EC Index Number:
202-723-9
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

≥98.0% (HPLC)

form

solid

autoignition temp.

500 °C

potency

7000 mg/kg LD50, oral (Rat)

pH

3.4 (20 °C in H2O, saturated solution)

bp

263 °C/1013 hPa

mp

108-111 °C

transition temp

flash point 150 °C

solubility

1 g/L

bulk density

500 kg/m3

storage temp.

2-30°C

SMILES string

OC(=O)c1cc(ccc1)C

InChI

1S/C8H8O2/c1-6-3-2-4-7(5-6)8(9)10/h2-5H,1H3,(H,9,10)

InChI key

GPSDUZXPYCFOSQ-UHFFFAOYSA-N

Application

  • Laccase-mediated Oligomerization: Explores the enzymatic oligomerization of 4-hydroxybenzoic acid which could lead to new materials with improved stability and functionality, useful in material science and drug delivery systems (Slagman et al., 2016).

Analysis Note

Assay (HPLC, area%): ≥ 98.0 % (a/a)
Melting range (lower value): ≥ 110 °C
Melting range (upper value): ≤ 113 °C
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

302.0 °F

Flash Point(C)

150 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service