Skip to Content
Merck
All Photos(1)

Key Documents

8.20365

Sigma-Aldrich

Cyclohexane-1,2-dicarboxylic anhydride

(mixture of isomeres) for synthesis

Synonym(s):

Cyclohexane-1,2-dicarboxylic anhydride, Hexahydrophthalic acid anhydride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H10O3
Molecular Weight:
154.16
MDL number:
UNSPSC Code:
12352115
EC Index Number:
236-086-3
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

0.01 hPa ( 20 °C)

Quality Level

form

powder

autoignition temp.

395 °C (DIN 51794)

potency

4040 mg/kg LD50, oral (Rat)

mp

30-33 °C

transition temp

flash point 152 °C

density

1.23 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O1C(=O)C2C(CCCC2)C1=O

InChI

1S/C8H10O3/c9-7-5-3-1-2-4-6(5)8(10)11-7/h5-6H,1-4H2

InChI key

MUTGBJKUEZFXGO-UHFFFAOYSA-N

Analysis Note

Assay (morpholine method): ≥ 98.0 %
Melting range (lower value): ≥ 30 °C
Melting range (upper value): ≤ 33 °C
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Pictograms

CorrosionHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service