8.14165
Iminodiacetic acid
≥96.0% (acidimetric), for peptide synthesis
Synonym(s):
Iminodiacetic acid
About This Item
Product Name
Iminodiacetic acid, for synthesis
Quality Level
Assay
≥96.0% (acidimetric)
form
crystals
potency
>2000 mg/kg LD50, oral (Rat)
pH
2.2-2.3 (20 °C in H2O, saturated solution)
mp
247 °C (decomposition)
solubility
42 g/L
storage temp.
2-30°C
SMILES string
N(CC(=O)O)CC(=O)O
InChI
1S/C4H7NO4/c6-3(7)1-5-2-4(8)9/h5H,1-2H2,(H,6,7)(H,8,9)
InChI key
NBZBKCUXIYYUSX-UHFFFAOYSA-N
Analysis Note
Identity (IR): passes test
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service