Skip to Content
Merck
All Photos(1)

Key Documents

8.09733

Sigma-Aldrich

1,2,3,4-Tetrahydronaphthalene

for synthesis

Synonym(s):

1,2,3,4-Tetrahydronaphthalene, Tetralin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12
CAS Number:
Molecular Weight:
132.20
MDL number:
UNSPSC Code:
12352005
EC Index Number:
204-340-2
NACRES:
NA.22
Assay:
≥98% (GC)
bp:
200-209 °C
vapor pressure:
0.34 hPa ( 20 °C)
Pricing and availability is not currently available.

vapor pressure

0.34 hPa ( 20 °C)

Quality Level

Assay

≥98% (GC)

form

liquid

autoignition temp.

425 °C (DIN 51794)

potency

2860 mg/kg LD50, oral (Rat)
16710 mg/kg LD50, skin (Rabbit)

expl. lim.

0.8-5 % (v/v)

bp

200-209 °C

mp

-30 °C

transition temp

flash point 71 °C

solubility

0.045 g/L

density

0.97 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

C1CCc2c(cccc2)C1

InChI

1S/C10H12/c1-2-6-10-8-4-3-7-9(10)5-1/h1-2,5-6H,3-4,7-8H2

InChI key

CXWXQJXEFPUFDZ-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.967 - 0.970
Identity (IR): passes test

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2

Supplementary Hazards

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

159.8 °F - closed cup

Flash Point(C)

71 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service