Skip to Content
Merck
All Photos(1)

Key Documents

8.08158

Sigma-Aldrich

N-Allylthiourea

for synthesis

Synonym(s):

N-Allylthiourea, Thiosinamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8N2S
CAS Number:
Molecular Weight:
116.18
MDL number:
UNSPSC Code:
12352111
EC Index Number:
203-683-5
NACRES:
NA.22

Quality Level

Assay

≥98.0% (HPLC)

form

powder

potency

200 mg/kg LD50, oral (Rat)

mp

70-73 °C

solubility

66 g/L

bulk density

550 kg/m3

storage temp.

2-30°C

SMILES string

S=C(NCC=C)N

InChI

1S/C4H8N2S/c1-2-3-6-4(5)7/h2H,1,3H2,(H3,5,6,7)

InChI key

HTKFORQRBXIQHD-UHFFFAOYSA-N

Application

  • Zn dendrite suppression and solid electrolyte interface control using N-allylthiourea as an electrolyte additive for aqueous Zn-ion batteries: The study highlights the effectiveness of N-allylthiourea as an additive for electrolytes in zinc-ion batteries to improve performance and safety (Source).
  • Design, synthesis, and spectral characterization of series of thiocarbamides with coumarin backbone: This research synthesizes a series of thiocarbamides including N-allylthiourea, exploring their spectral properties and potential pharmaceutical applications (Source).

Analysis Note

Assay (HPLC, area%): ≥ 98.0 % (a/a)
Identity (IR): passes test

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service