Skip to Content
Merck
All Photos(1)

Key Documents

8.04146

Sigma-Aldrich

4-Bromobenzaldehyde

for synthesis

Synonym(s):

4-Bromobenzaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
4-(Br)C6H4CHO
CAS Number:
Molecular Weight:
185.02
MDL number:
UNSPSC Code:
12352114
EC Index Number:
214-365-0
NACRES:
NA.22

Quality Level

form

powder

bp

123-125 °C/31 hPa

mp

55-58 °C

transition temp

flash point 108 °C

density

1.85 g/cm3

bulk density

670 kg/m3

storage temp.

2-30°C

SMILES string

Brc1ccc(cc1)C=O

InChI

1S/C7H5BrO/c8-7-3-1-6(5-9)2-4-7/h1-5H

InChI key

ZRYZBQLXDKPBDU-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

228.2 °F - closed cup

Flash Point(C)

109 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service