Skip to Content
Merck
All Photos(1)

Key Documents

7110-OP

Sigma-Aldrich

Phenylmethyl Sulfonyl Fluoride

OmniPur® Grade, >=99.0%

Synonym(s):

OmniPur® Phenylmethyl Sulfonyl Fluoride, PMSF

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥99%

form

crystals (needle-like)

mol wt

174.19 g/mol

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

shipped in

ambient

SMILES string

F[S](=O)(=O)Cc1ccccc1

InChI

1S/C7H7FO2S/c8-11(9,10)6-7-4-2-1-3-5-7/h1-5H,6H2

InChI key

YBYRMVIVWMBXKQ-UHFFFAOYSA-N

General description

Molecular Biology Grade.

Assay: ≥99.0%
Melting point: 91-95°C





Intended for laboratory and manufacturing use only. Not for drug, food, or household use.

Warning

Toxicity: Corrosive (L)

Other Notes

Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
OMNIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jessica A Espino et al.
Journal of visualized experiments : JoVE, (158) (2020-04-21)
Fast oxidation of proteins (FPOP) is a hydroxyl radical protein footprinting (HRPF) method used to study protein structure, protein-ligand interactions, and protein-protein interactions. FPOP utilizes a KrF excimer laser at 248 nm for photolysis of hydrogen peroxide to generate hydroxyl
Feng Yue et al.
Cell reports, 38(3), 110267-110267 (2022-01-20)
The lipid droplet (LD) is a central hub for fatty acid metabolism in cells. Here we define the dynamics and explore the role of LDs in skeletal muscle satellite cells (SCs), a stem cell population responsible for muscle regeneration. In
Kun Ho Kim et al.
Cell reports, 42(11), 113329-113329 (2023-10-26)
Development is regulated by various factors, including protein methylation status. While PRMT5 is well known for its roles in oncogenesis by mediating symmetric di-methylation of arginine, its role in normal development remains elusive. Using Myod1Cre to drive Prmt5 knockout in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service