Skip to Content
Merck
All Photos(2)

Key Documents

551850

Sigma-Aldrich

Quinacrine dihydrochloride

A non-specific phospholipase A2 (PLA2) inhibitor.

Synonym(s):

Mepacrine, 6-Chloro-9-(4-diethylamino-1-methylbutylamino)-2-methoxyacridine dihydrochloride, Atebrin dihydrochloride, Mepacrine dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H30ClN3O · 2HCl
CAS Number:
Molecular Weight:
472.88
Beilstein:
4834013
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

description

Merck USA index - 14, 8044

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

yellow

solubility

water: soluble

shipped in

ambient

storage temp.

10-30°C

SMILES string

Cl[H].Cl[H].CCN(CC)CCCC(C)Nc1c2ccc(Cl)cc2nc3ccc(OC)cc13

InChI

1S/C23H30ClN3O.2ClH/c1-5-27(6-2)13-7-8-16(3)25-23-19-11-9-17(24)14-22(19)26-21-12-10-18(28-4)15-20(21)23;;/h9-12,14-16H,5-8,13H2,1-4H3,(H,25,26);2*1H

InChI key

UDKVBVICMUEIKS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

A non-specific phospholipase A2 (PLA2) inhibitor. Acts as an acetylcholine receptor antagonist. Suppresses glibenclamide-sensitive K+-currents (IC50 = 4.4 µM). Also inhibits monoamine oxidase (MAO).

Biochem/physiol Actions

Cell permeable: no
Non-selective MAO-A/B inhibitor.
Product does not compete with ATP.
Reversible: no
Target IC50: 4.4 µM in suppressing glibenclamide-sensitive K+-currents

Warning

Toxicity: Harmful (C)

Reconstitution

Stock solutions are stable for up to 60 h at 20°C.

Other Notes

Holscher, C. 1995. Neurobiol. Learn. Mem. 63, 206.
Sakuta, H. and Yoneda, I. 1994. Eur. J. Pharmacol. 252, 117.
Sargent, C.A., et al 1992. J. Pharmacol. Exp. Ther.262, 1161.
Voorhis, B.J., et al. 1992. Invertebr. Reprod. Dev.21, 33.
Murakami, M., et al. 1991. Biochem. Biophys. Res. Commun.181, 714.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service