Skip to Content
Merck
All Photos(1)

Key Documents

5.30032

Sigma-Aldrich

Nedd4/Rsp5 Signaling Inducer, NAB2

Synonym(s):

Nedd4/Rsp5 Signaling Inducer, NAB2, N-(2-Chlorobenzyl)-1-(2,5-dimethylphenyl)-1H-benzo[d]imidazole-5-carboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H20ClN3O
CAS Number:
Molecular Weight:
389.88
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 100 mg/mL

storage temp.

2-8°C

SMILES string

CC1=CC(=C(C=C1)C)N2C=NC3=C2C=CC(=C3)C(=O)NCC4=CC=CC=C4Cl

InChI key

CZSLEMCYYGEGKP-UHFFFAOYSA-N

General description

A cell-permeable N-aryl benzimidazole (NAB) compound that protects against α-synuclein (α-syn) toxicity in yeast (0.625 to 10 µM), wt human α-syn-expressing C. elegans dopaminergic (DA) neurons (10 µM), human A53T α-syn-expressing rat DA neurons (0.1 µM), as well as in human neurons differentiated from iPSCs derived from Parkinson′s disease (PD) patients carrying either AT53T α-syn or an α-syn triplication (5 to 40 µM) by restoring Nedd4 (mammalian)/Rsp5 (yeast) E3 ligase-dependent processes, including ER-to-Golgi, plasma membrane- and Golgi-to-vacuole/lysosome trafficking events, reducing α-syn-caused nitrosative and ER stress. NAB2 is also reported to inhibit the growth, but not viability, of non-α-syn-expressing yeast via the same mechanism of action (MOA) albeit at a lower potency (GI40 = 20.5 µM vs. EC40 = 4.5 µM against α-syn toxicity). Although the exact molecular mechanism is yet unknown, NAB2 does not affect cellular α-syn level or Nedd4/Rsp5 ligase activity.
A cell-permeable N-aryl benzimidazole (NAB) compound that protects against α-synuclein (α-syn) toxicity in yeast (0.625 to 10 µM), wt human α-syn-expressing C. elegans dopaminergic neurons (10 µM), human A53T α-syn-expressing rat DA neurons (0.1 µM), as well as in Parkinson′s disease patients-derived neurons carrying AT53T α-syn or an α-syn triplication (5 to 40 µM) by restoring Nedd4 (mammalian)/Rsp5 (yeast) E3 ligase-dependent trafficking events, reducing α-syn-caused nitrosative and ER stress without affecting cellular α-syn level or Nedd4/Rsp5 ligase activity.
A cell-permeable compound that protects against alpha-synuclein toxicity.

Biochem/physiol Actions

Cell permeable: yes

Caution

NAB2 is moderately hydrophobic (clogP ~5.2)and can precipitate out of solution if added directly to cell culture or yeast media. Care should therefore be taken to optimize solubility and guard against false negative results caused by irrecoverable precipitation of the compound. Please refer to the Recommended Reaction Conditions above for details.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Tardiff, D.F., et al. 2013. Science342, 979.
Chung, C.Y., et al. 2013. Science342, 983.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service