Skip to Content
Merck
All Photos(1)

Key Documents

331569

Sigma-Aldrich

Ethidium Bromide Adsorber

Developed specifically for the safe and simple removal of ethidium bromide (EtBr) from aqueous staining solutions and running buffers used in nucleic acid separation gels.

Synonym(s):

Ethidium Bromide Adsorber

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.25

Quality Level

form

suspension

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

gray-brown

shipped in

ambient

storage temp.

15-25°C

SMILES string

C(C)c3c(cccc3)C=C.c2(c(cccc2)C=C)C=C.c1(ccccc1)C=C

InChI

1S/C10H12.C10H10.C8H8/c2*1-3-9-7-5-6-8-10(9)4-2;1-2-8-6-4-3-5-7-8/h3,5-8H,1,4H2,2H3;3-8H,1-2H2;2-7H,1H2

InChI key

NWUYHJFMYQTDRP-UHFFFAOYSA-N

General description

Developed specifically for the safe and simple removal of ethidium bromide (EtBr) from aqueous staining solutions and running buffers used in nucleic acid separation gels. Typical concentrations in these solutions are 0.5 - 10 mg/L. EtBr is bound and concentrated by the adsorber column, allowing removal of the filtrate. Each column normally binds at least 300 mg of ethidium bromide from TAE or TBE buffers. Residual capacity is easily visualized because the EtBr appears as a dark red to black band on the column. As long as this band has not reached the bottom of the column bed, the concentration of dye is still below the column capacity.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Removal of ethidium bromide (EtBr) from aqueous staining solutions and running buffers
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Irritant (B)

Reconstitution

When the adsorber column is not in use, both ends should be closed off. The column should not be allowed to dry out, as air bubbles may form in the column, hindering flow rate.

Other Notes

MacGregor, J.T., and Johnson, I.J. 1977. Mutat. Res.48, 103.
Waring, M. 1975. In: Antibiotics, Corcoran, J.W., and Hahn, F.E., Editors, Vol. 3, 141, Cold Spring Harbor, NY.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service