Skip to Content
Merck
All Photos(1)

Key Documents

1.17290

Millipore

Profinet Module

for use with MAS-100® Iso

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.74

feature

electronic

application(s)

air monitoring
pharmaceutical

compatibility

for use with MAS-100® Iso

General description

Profinet Module for MAS-100 ISO® NT/MH with Profibus interface is designed as an accessory which can be used to convert Profibus to Profinet communication. The accessory is pre-configured to be used by Siemens integrators with access to the TIA portal and allows to operate the MAS-100 ISO® NT/MH instruments with Siemens S7 PLC′s.

Application

Profinet Module for MAS-100 ISO® NT/MH with Profibus is an accessory system that can be used with MAS-100 ISO® NT/MH microbial air samplers to convert Profibus to Profinet communication.

Features and Benefits

The new Profinet Module for MAS-100 ISO® NT/MH instrument has the following features:
  • used as a Profibus Master.
  • as Profinet Client.
  • Converts the data.

Components

The key component and pre-requisites of Profinet Module is:
  • Control Unit with Profibus Main Road (pre-configured with file: MBV_063C.gsd)
  • Integrator requires license for TIA portal at Siemens.

Legal Information

MAS-100 is a registered trademark of MBV AG
MAS-100 ISO is a registered trademark of MBV AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service