Skip to Content
Merck
All Photos(2)

Key Documents

1.16732

Supelco

Nitrite Test Strips, reflectometric

0.03-1.00 g/L (NO₂⁻), REFLECTOQUANT®, for use with RQflex® 20

Synonym(s):

Nitrite chemical test strips, Nitrite test strip for instrument reader

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24
Pricing and availability is not currently available.

Product Name

Nitrite Test, reflectometric, 0.03-1.00 g/L (NO2-), for use with REFLECTOQUANT®

description

quantitative

Quality Level

specific analyte(s)

nitrite

reaction suitability

reactivity: amine reactive

measuring range

0.03-1.00 g/L (NO2-)

compatibility

RQflex® 20
for use with REFLECTOQUANT®

detection method

reflectometric

storage temp.

2-8°C

General description

Method: reflectometric with test strips 0.03 - 1.00 g/l NO₂⁻ Reflectoquant®
Nitrite is naturally present in soil, water and food, and regularly testing its concentration is essential for many applications. Nitrites must be closely monitored in drinking and groundwater as well as in process, waste, or aquarium water. In farm produce and fruit juices, high nitrate concentrations affect quality and keeping properties and can constitute serious health risks. Nitrites are also used in additives for food products and serve as corrosion inhibitors in engine cooling systems. On the Reflectoquant Nitrite test strips, nitrite ions react with an aromatic amine to form an orange-red azo dye that is determined reflectometrically on the RQflex® 20. For pre-testing a sample in an economic and quick way to estimate the nitrite concentration and a possible dilution factor prior to using this quantitative method we recommend our semi-quantitative MQuant test strips for nitrite. Further quantitative determination methods can be found with our Spectroquant tests.

Analysis Note

function control: conforms
Assay
- standard 1:
0.09 - 0.11 g/l
- standard 2: 0.45 - 0.55 g/l
- standard 3: 0.72 - 0.88 g/l

Legal Information

REFLECTOQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
RQFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service