Skip to Content
Merck
All Photos(3)

Key Documents

1.07025

Supelco

Tetrahydrofuran

≥99.5% (GC), EMPARTA® ACS reagent

Synonym(s):

THF, Tetramethylene oxide, Oxolane, Butylene oxide, Butylene oxide, Oxolane, Tetramethylene oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8O
CAS Number:
Molecular Weight:
72.11
Beilstein:
102391
MDL number:
UNSPSC Code:
12191504
EC Index Number:
203-726-8
NACRES:
NB.21
grade:
ACS reagent
bp:
65-67 °C (lit.)
vapor pressure:
114 mmHg ( 15 °C)
143 mmHg ( 20 °C)

Product Name

Tetrahydrofuran, for analysis EMPARTA® ACS

grade

ACS reagent

Quality Level

vapor density

2.5 (vs air)

vapor pressure

114 mmHg ( 15 °C)
143 mmHg ( 20 °C)

product line

EMPARTA®

Assay

≥99.5% (GC)

form

liquid

autoignition temp.

610 °F

expl. lim.

1.8-11.8 %

dilution

(for routine analytical applications)

impurities

≤0.01% Peroxide (as H2O2)
≤0.05% Water

evapn. residue

≤0.03%

color

clearAPHA: ≤10

refractive index

n20/D 1.407 (lit.)

pH

7-8 (20 °C, 200 g/L in H2O)

bp

65-67 °C (lit.)

mp

−108 °C (lit.)

transition temp

flash point -21.5 °C

density

0.889 g/mL at 25 °C (lit.)

storage temp.

2-30°C

SMILES string

C1CCOC1

InChI

1S/C4H8O/c1-2-4-5-3-1/h1-4H2

InChI key

WYURNTSHIVDZCO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

EMD Millipore offers EMPARTA grade solvents for routine tasks in analytical laboratories. EMPARTA grade solvents offer fewer test parameters than EMSURE, yet are still of high quality. EMPARTA solvents meet ACS requirements, making them ideal for a wide range of analytical applications.

Application



  • Phenylalanine Ammonia Lyase Activity Research: A study investigated the mutase-like activity of phenylalanine ammonia lyase from Anabaena variabilis, demonstrating the utility of buffer concentrate in biochemical assays to stabilize enzyme activity during the research (Weise et al., 2018).


  • Studying Amyloid Precursor Protein: A study used buffer concentrate for experiments involving the amyloid precursor protein C-terminal fragment C100, noting its importance in maintaining the stability of protein conformations during interactions with γ-secretase modulators (Botev et al., 2011).




Analysis Note

Purity (GC): ≥ 99.5 %
Identity (IR): conforms
Appearance: clear
Color: ≤ 10 Hazen
Peroxide (as H₂O₂): ≤ 0.01 %
Evaporation residue: ≤ 0.03 %
Water: ≤ 0.05 %
Stabilized with 2,6-Di-tert-butyl-4-methylphenol (BHT).

Legal Information

EMPARTA is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-6.2 °F - closed cup

Flash Point(C)

-21.2 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service