Skip to Content
Merck
All Photos(1)

Key Documents

1.00861

Supelco

Zinc Test, photometric

photometric, 0.025-1.000 mg/L (Zn), Spectroquant®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

product name

Zinc Cell Test, photometric, 0.025-1.000 mg/L (Zn), Spectroquant®

product line

Spectroquant®

Quality Level

usage

sufficient for 25 tests

specific analyte(s)

zinc

measuring range

0.025-1.000 mg/L (Zn)

technique(s)

photometry: suitable

compatibility

for use with Spectroquant® Move 100
for use with Spectroquant® Nova 60 A
for use with Spectroquant® Prove 100
for use with Spectroquant® Prove 300
for use with Spectroquant® Prove 600

detection method

photometric (PAR)

storage temp.

15-25°C

General description

Method: photometric 0.025 - 1.000 mg/l Zn Spectroquant®

This Spectroquant® Zinc test allows the accurate quantification of zinc ions in various waters. In alkaline solution zinc ions react with pyridylazoresorcinol (PAR) to form a red complex that is determined photometrically.
MQuant® test strips can be used for pre-testing a sample in an economic and quick way to estimate the zinc concentration and a possible dilution factor prior to using a quantitative method such as the Spectroquant® test.
All our Cell and Reagent Test Kits are equipped with the unique Live ID (2D barcode) which allows seamless method recognition and contains essential information such as lot number, expiry date, and automatic calibration updates.
The Spectroquant® Cell Tests come with prefilled 16 mm round cells as well as all the required reagents to perform the analysis according to the procedure described in the accompanying instruction leaflet.

Legal Information

MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Oral - Acute Tox. 3 Inhalation - Acute Tox. 4 Dermal - Aquatic Chronic 3 - Carc. 1B - Eye Dam. 1 - Muta. 2 - Skin Corr. 1 - Skin Sens. 1 - STOT RE 2 - STOT SE 2 - STOT SE 3

Target Organs

Respiratory system, Thyroid

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Photometric determination with pyridylazoresorcinol (PAR) subsequent to acid extraction

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service