Skip to Content
Merck
All Photos(1)

Documents

1.00223

Sigma-Aldrich

Methyl benzoate

(distilled and stabilized Type A), Technipur®

Synonym(s):

Methyl benzenecarboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5COOCH3
CAS Number:
Molecular Weight:
136.15
Beilstein:
1072099
MDL number:
UNSPSC Code:
12352100
NACRES:
NA.05

vapor density

4.68 (vs air)

Quality Level

vapor pressure

<1 mmHg ( 20 °C)
0.36 hPa ( 20 °C)

refractive index

n20/D 1.516 (lit.)

bp

198-199 °C (lit.)

mp

−12 °C (lit.)

density

1.088 g/mL at 20 °C (lit.)

SMILES string

COC(=O)c1ccccc1

InChI

1S/C8H8O2/c1-10-8(9)7-5-3-2-4-6-7/h2-6H,1H3

InChI key

QPJVMBTYPHYUOC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methyl benzoate is a volatile aromatic ester widely used in perfumery industries because of its pleasant smell. It is also an important class of chemical intermediate commonly used in plasticizers, pharmaceuticals, dyeing polyester fibers, solvents, disinfectants, and pesticides.

Application

Methyl benzoate can be used as:
  • A solvent in the ink-jet printing of polymers.
  • A nonreacting plasticizer in the efficient crosslinking reaction of phenolic resin.
  • A starting material to synthesize glycol dibenzoate plasticizers by transesterification with glycols.

Legal Information

Technipur is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

170.6 °F - Pensky-Martens closed cup

Flash Point(C)

77 °C - Pensky-Martens closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The use of thermogravimetry to follow the rate of evaporation of an ingredient used in perfumes
Aggarwal P, et al.
Journal of Thermal Analysis and Calorimetry, 49(2), 595-599 (1997)
An analysis of the role of nonreactive plasticizers in the crosslinking reactions of a rigid resin
Patel JP, et al.
Journal of Polymer Science. Part B, Polymer Physics, 55(2), 206-213 (2017)
Influence of the glycol component in dibenzoate plasticizers on the properties of plasticized PVC films
Wardzinska E and Penczek P
Journal of Applied Polymer Science, 97(3), 822-824 (2005)
Ink-jet printing of polymers-from single dots to thin film libraries
Tekin E, et al.
Journal of Materials Chemistry, 14(17), 2627-2632 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service