Skip to Content
Merck
All Photos(1)

Key Documents

880320P

Avanti

18:0 PEG3000 PE

Avanti Research - A Croda Brand 880320P, powder

Synonym(s):

1,2-distearoyl-sn-glycero-3-phosphoethanolamine-N-[methoxy(polyethylene glycol)-3000] (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 25 mg (880320P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 880320P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCCNC(OCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOC)=O)=O)(OC(CCCCCCCCCCCCCCCC

General description

18:0 PEG3000 PE or 1,2-distearoyl-sn-glycero-3-phosphoethanolamine-N-[methoxy(polyethylene glycol)-3000] is a PEGylated lipopolymer.

Application

18:0 PEG3000 PE or 1,2-distearoyl-sn-glycero-3-phosphoethanolamine-N-[methoxy(polyethylene glycol)-3000] is suitable for fabricating non-supported sensor like quartz crystal microbalance (QCM) odor sensors and in the preparation of holmium nanoparticles.

Packaging

5 mL Amber Glass Screw Cap Vial (880320P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anthony J Di Pasqua et al.
Small (Weinheim an der Bergstrasse, Germany), 8(7), 997-1000 (2012-02-03)
Nanoparticles containing stable holmium ((165) Ho) are prepared by nanotemplate engineering and subsequently irradiated in a neutron flux to yield (166) Ho, a beta-emitting radiotherapeutic isotope. After intraperitoneal injection to mice bearing SKOV-3 ovarian tumors, significant tumor accumulation of the
Chang Hyun Kim et al.
International journal of pharmaceutics, 583, 119393-119393 (2020-05-08)
Docetaxel (DTX) has poor solubility, low specificity, and severe side effects. For efficient targeting of DTX to hepsin-overexpressing SKOV3 ovarian cancer cells, PEGylated and RIPL peptide (IPLVVPLRRRRRRRRC)-conjugated nanostructured lipid carriers (PEG-RIPL-NLCs) were examined for in vitro and in vivo antitumor
Chemisorbed PEGylated lipopolymers as sensing film supports for QCM odor sensors
Wyszynski B, et al.
Sensors and Actuators B, Chemical, 130(2), 857-863 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service