Skip to Content
Merck
All Photos(1)

Key Documents

870726P

Avanti

26:0 Coenzyme A

Avanti Research - A Croda Brand 870726P, powder

Synonym(s):

hexacosanoyl Coenzyme A (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C47H95N10O17P3S
CAS Number:
Molecular Weight:
1197.30
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 10 mg (870726P-10mg)
pkg of 1 × 5 mg (870726P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 870726P

application(s)

lipidomics

lipid type

coenzymes

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@@](C(NCCC(NCCSC(CCCCCCCCCCCCCCCCCCCCCCCCC)=O)=O)=O)(C(C)(COP([O-])(OP([O-])(OC[C@H]([C@H]1OP([O-])(O)=O)O[C@H]([C@@H]1O)N2C3=C(C(N)=NC=N3)N=C2)=O)=O)C)[H].[NH4+].[NH4+].[NH4+]

InChI

1S/C47H86N7O17P3S.3H3N/c1-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23-24-25-26-27-38(56)75-31-30-49-37(55)28-29-50-45(59)42(58)47(2,3)33-68-74(65,66)71-73(63,64)67-32-36-41(70-72(60,61)62)40(57)46(69-36)54-35-53-39-43(48)51-34-52-44(39)54;;;/h34-36,40-42,46,57-58H,4-33H2,1-3H3,(H,49,55)(H,50,59)(H,63,64)(H,65,66)(H2,48,51,52)(H2,60,61,62);3*1H3/t36-,40?,41+,42+,46-;;;/m1.../s1

InChI key

XKPNJROJQZVENY-PQBGVFARSA-N

General description

26:0 Coenzyme A , also known as hexacosanoyl Coenzyme A, is a coenzyme derivative of hexacosanoic acid. It is found in high concentration in ATP binding cassette transporter D1-deficient cells.

Application

26:0 Coenzyme A has been used as a substrate for microsomal enzyme 1-acylglycerol-3-phosphate-O-acyltransferase 11 (AGPAT11).

Packaging

5 mL Amber Glass Screw Cap Vial (870726P-10mg)
5 mL Amber Glass Screw Cap Vial (870726P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anil K Agarwal et al.
Journal of lipid research, 51(8), 2143-2152 (2010-04-07)
The conversion of lysophosphatidic acid (LPA) to phosphatidic acid is carried out by the microsomal enzymes 1-acylglycerol-3-phosphate-O-acyltransferases (AGPATs). These enzymes are specific for acylating LPA at the sn-2 (carbon 2) position on the glycerol backbone and are important, because they
Oshrit Ben-David et al.
The Journal of biological chemistry, 286(34), 30022-30033 (2011-06-28)
Sphingolipids (SLs) act as signaling molecules and as structural components in both neuronal cells and myelin. We now characterize the biochemical, histological, and behavioral abnormalities in the brain of a mouse lacking very long acyl (C22-C24) chain SLs. This mouse
Kotaro Hama et al.
Journal of lipid research, 61(4), 523-536 (2020-02-23)
X-linked adrenoleukodystrophy (X-ALD) is an inherited disorder caused by deleterious mutations in the ABCD1 gene. The ABCD1 protein transports very long-chain FAs (VLCFAs) from the cytosol into the peroxisome where the VLCFAs are degraded through β-oxidation. ABCD1 dysfunction leads to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service