Skip to Content
Merck
All Photos(2)

Documents

850305C

Avanti

06:0 PC (DHPC)

1,2-dihexanoyl-sn-glycero-3-phosphocholine, chloroform

Synonym(s):

DHPC; 1,2-dicaproyl-sn-glycero-3-phosphocholine; PC(6:0/6:0)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H40NO8P
CAS Number:
Molecular Weight:
453.51
UNSPSC Code:
51191904
NACRES:
NA.25

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (850305C-25mg)
pkg of 2 × 20 mL (850305C-1g)
pkg of 2 × 4 mL (850305C-200mg)

manufacturer/tradename

Avanti Research - A Croda Brand 850305C

concentration

10 mg/mL (850305C-25mg)
25 mg/mL (850305C-1g)
25 mg/mL (850305C-200mg)

lipid type

cardiolipins
phospholipids

shipped in

dry ice

storage temp.

−20°C

InChI

1S/C20H40NO8P/c1-6-8-10-12-19(22)26-16-18(29-20(23)13-11-9-7-2)17-28-30(24,25)27-15-14-21(3,4)5/h18H,6-17H2,1-5H3

InChI key

DVZARZBAWHITHR-UHFFFAOYSA-N

General description

Phosphatidylcholines (PCs) are phospholipids, that are abundantly present in the tissues of animal. PCs are known to carry saturated and unsaturated fatty acids in the sn-1 and sn-2 position respectively.
The list of Phosphatidylcholine products offered by Avanti is designed to provide compounds having a variety of physical properties. Products available include short chain (C3-C8 are water soluble and hygroscopic), saturated, multi-unsaturated and mixed acid PC′s. All of the products are purified by HPLC, and special precautions are taken to protect the products for oxidization and hydrolysis.

Application

06:0 PC (1,2-dihexanoyl-sn-glycero-3-phosphocholine/DHPC) may be used:
  • in the preparation of lipid bicelles for nuclear magnetic resonance (NMR) experiments
  • to form lipid vesicles/micelles for guanosine triphosphate (GTP) hydrolysis (GTPase) assays
  • in the reconstitution of p7 protein of the hepatitis C virus (p7(5a)) in bicelles

Biochem/physiol Actions

Phosphatidylcholines (PCs) plays a functional role in reducing the level of blood lipid and cholesterol. It may also have the ability to inhibit cardiovascular disease and obesity.

Packaging

30 mL Amber Narrow Mouth Glass Bottle with Screw Cap (850305C-1g)
5 mL Clear Glass Sealed Ampule (850305C-200mg)
5 mL Clear Glass Sealed Ampule (850305C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Lipid binding by disordered proteins
Perez Y, et al.
Protocol Exchange, 10 null
Lipids II: Phospholipids, Glycosphingolipids, and Cholesterol
Bhagavan NV, et al.
Essentials of Medical Biochemistry, 299-320 null
Mutagenesis and peptide analysis of the DRY motif in the alpha 2A adrenergic receptor: evidence for alternate mechanisms in G protein-coupled receptors
Chung DA, et al.
Biochemical and biophysical research communications, 293(4), 1233-1241 null
Gabriel A Cook et al.
Biochemistry, 52(31), 5295-5303 (2013-07-12)
Hepatitis C virus (HCV) protein p7 plays an important role in the assembly and release of mature virus particles. This small 63-residue membrane protein has been shown to induce channel activity, which may contribute to its functions. p7 is highly
Qinghai Zhang et al.
Journal of the American Chemical Society, 130(23), 7357-7363 (2008-05-16)
The rate limiting step in biophysical characterization of membrane proteins is often the availability of suitable amounts of protein material. It was therefore of interest to demonstrate that microcoil nuclear magnetic resonance (NMR) technology can be used to screen microscale

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service