Z742018
Duran® Bottle Tags GL 45
Eight colors: two bottle tags are supplied with each color
Synonym(s):
292432904
All Photos(2)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.17
Recommended Products
packaging
pack of 16
manufacturer/tradename
DURAN® 292432904
Related Categories
General description
- Colourful silicone GL 45 Bottle Tags can be easily and securely attached around the neck of the YOUTILITY bottle
- Distinct colours for easy customisation and unambiguous bottle identification
- Elastic bottle tags will fit around any DIN GL 45 bottle neck
- Can be applied with the screw cap in place
- From silicone, eight colours, pack of 16
Features and Benefits
- Available in different colours for identification marking and personalizing laboratory bottles
- The elastic Bottle Tags can be easily and securely attached around the neck, even with the screw cap in place
- Feature a graspable tab that makes them easy to remove with bare or gloved hands
- Chemically robust, heat resistant and totally reusable
- Fully compatible with all the GL 45 threads and therefore highly versatile
Legal Information
DURAN is a registered trademark of DWK Life Sciences
Choose from one of the most recent versions:
Certificates of Analysis (COA)