Key Documents
Z286486
Waste solvent funnels
for container cap. 4 L,
closure size 38 - 430
Synonym(s):
Solvent Funnels, Waste Funnels
About This Item
UNSPSC Code:
41121809
NACRES:
NB.43
Pricing and availability is not currently available.
funnel diam.
8 in.
Related Categories
General description
ECO Funnels prevent waste solvent evaporation. We recommend that the ECO Funnel be removed from the waste container at the end of each day or when it is not in use for long periods of time.
Specifications:
  • Chemically resistant HDPE and PP construction
  • Lip-seal on lid (red) seals the waste system when closed
  • Funnel stem sealed to waste container cap prevents emission from sides of stem
  • Pressure-equalizing tube prevents solvent trapping
Funnels are sized to fit the following waste solvent containers:
  • 4 L funnel fits round bottle, B8410
  • 8 L funnel fits round bottle, B8535
  • 9 L funnel fits rectangular carboy, C6317
  • 20 L funnel fits rectangular D.O.T. carboy, Z268208
Order funnel and waste container separately.
Choose from one of the most recent versions:
Certificates of Analysis (COA)