Z249963
Wheaton narrow mouth bottles with caps
capacity 8 oz (250 mL), amber, black plastic cap, Poly-Seal® cone liner
Synonym(s):
Wheaton bottle, Wheaton bottles, narrow mouth
About This Item
Recommended Products
material
amber bottle
black plastic cap
glass bottle
round bottle
Joint
Joints threaded neck (24-400)
feature
with cap
packaging
pkg of 12 ea
manufacturer/tradename
WHEATON® W216851
capacity
8 oz (250 mL)
diam. × H
60 mm × 136 mm
color
amber
Looking for similar products? Visit Product Comparison Guide
General description
Choice of cap styles:
- White PP cap with PTFE or polyvinyl liner
- Black plastic cap with Poly-Seal® PE cone liner
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service