Key Documents
Z167576
Sigma-Aldrich
Aldrich® Schlenk-type hex head stopper
male joint: ST/NS 14/20
About This Item
UNSPSC Code:
41122500
NACRES:
NB.43
Pricing and availability is not currently available.
label
M
Joint
male joint: ST/NS14/20
General description
Thermoplastic screw caps with Viton O-rings usable to 140°C. This stopper has a hexagonal top.
Linkage
See the Threaded Ground Glass Tutorial to see how threaded joints work.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Articles
Versatile system for chemical reactions, extractions, and transfers of solids and liquids in laboratories.
Versatile system for chemical reactions, extractions, and transfers of solids and liquids in laboratories.
Versatile system for chemical reactions, extractions, and transfers of solids and liquids in laboratories.
Versatile system for chemical reactions, extractions, and transfers of solids and liquids in laboratories.