Key Documents
Z115533
Scienceware® protective lab-labeling system labels
size 1 1/2 in. × 3 in.
About This Item
UNSPSC Code:
55121611
NACRES:
NB.35
Pricing and availability is not currently available.
material
paper
white
packaging
pkg of 10 (100 labels per pad)
manufacturer/tradename
Bel-Art F13455-0015
size
1 1/2 in. × 3 in.
color
white
General description
Adhesive is resistant to casual exposure to acids, alkalis, and organic solvents. Temp range: -196 to 120 °C.
For a waterproof covering, purchase protective lab-labeling system tape to protect these labels.
Legal Information
Scienceware is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)