Key Documents
Z100900
Double-tipped needle
gauge 16, L 36 in.
About This Item
UNSPSC Code:
41122002
NACRES:
NB.03
Pricing and availability is not currently available.
packaging
pack of 1 ea
manufacturer/tradename
Cadence Science
L
36 in.
gauge
16
General description
304 SS with deflected tips.
Choose from one of the most recent versions:
Certificates of Analysis (COA)