Skip to Content
Merck
All Photos(1)

Documents

W509647

Sigma-Aldrich

(−)-Caryophyllene oxide

95%

Synonym(s):

β-Caryophyllene epoxide, (−)-Epoxycaryophyllene, (−)-Epoxydihydrocaryophyllene, (1R,4R,6R,10S)-9-Methylene-4,12,12-trimethyl-5-oxatricyclo[8.2.0.04,6]dodecane, trans-Caryophyllene oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H24O
CAS Number:
Molecular Weight:
220.35
FEMA Number:
4085
Beilstein:
148213
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
16.043
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

Fragrance grade

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009

Assay

95%

optical activity

[α]20/D −70°, c = 2 in chloroform

mp

62-63 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

β-caryophyllene (ox.)

Organoleptic

woody

SMILES string

[H][C@@]12CCC(=C)[C@@]3([H])CC(C)(C)[C@]3([H])CC[C@@]1(C)O2

InChI

1S/C15H24O/c1-10-5-6-13-15(4,16-13)8-7-12-11(10)9-14(12,2)3/h11-13H,1,5-9H2,2-4H3/t11-,12-,13-,15-/m1/s1

InChI key

NVEQFIOZRFFVFW-RGCMKSIDSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Synergistic Antinociceptive Effect of β-Caryophyllene Oxide in Combination with Paracetamol, and the Corresponding Gastroprotective Activity.: This study reveals that β-Caryophyllene oxide enhances the pain-relieving effects of paracetamol while providing gastroprotective benefits, suggesting its potential in pain management therapies (Espinosa-Juárez et al., 2024).


Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Muhammad Iqbal Choudhary et al.
Journal of natural products, 69(10), 1429-1434 (2006-10-28)
Microbial transformation of the sesquiterpene (-)-caryophyllene oxide (1) [(1R,4R,5R,9S)-4,5-epoxycaryophyllan-8(13)-ene] by a number of fungi, using a standard two-stage fermentation technique, has afforded as products (1R,4R,5R,9S)-4,5-dihydroxycaryophyllan-8(13)-ene (2), (1S,4R,5R,8S,9S)-clovane-5,9-diol (3), (1R,4R,5R,9S,11R)-4,5-epoxycaryophyllan-8(13)-en-15-ol (4), (1R,4R,5R,9S,11S)-4,5-epoxycaryophyllan-8(13)-en-14-ol (5), (1R,2S,4R,5R,9S)-4,5-epoxy-13-norcaryophyllan-8-one (6), (1R,4R,5R,8S,9S)-4,5-epoxycaryophyllan-13-ol (7), (1R,4R,5R,8S, 9S,13S)-caryolane-5,8,13-triol (8), (1R,3R,4R,5R,8S,9S)-4,5-epoxycaryophyllan-3,13-diol
M Salomé Gachet et al.
Journal of natural products, 74(4), 559-566 (2011-03-29)
In a survey of plants from Ecuador with antiprotozoal activity, Cupania cinerea was found to show significant in vitro activity against the Plasmodium falciparum K1 strain and Trypanosoma brucei rhodesiense. Subsequently, activity-guided isolation of the n-hexane and dichloromethane extracts from
Fangfang Zeng et al.
Insect biochemistry and molecular biology, 113, 103213-103213 (2019-08-24)
Mosquitoes rely heavily on the olfactory system to find a host for a bloodmeal, plants for a source of energy and suitable sites for oviposition. Here, we examined a cluster of eight odorant receptors (ORs), which includes one OR, CquiOR1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service