Skip to Content
Merck
All Photos(1)

Key Documents

W348600

Sigma-Aldrich

Ethyl trans-2-butenoate

≥96%, FG

Synonym(s):

Ethyl (E)-2-crotonate, Ethyl crotonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH=CHCOOC2H5
CAS Number:
FEMA Number:
3486
EC Number:
Council of Europe no.:
2244
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.248
NACRES:
NA.21
grade:
FG
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 172.515

Assay

≥96%

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

caramel; pungent; rum; sweet

SMILES string

O(CC)C(=O)\C=C\C

InChI

1S/C6H10O2/c1-3-5-6(7)8-4-2/h3,5H,4H2,1-2H3/b5-3+

InChI key

ZFDIRQKJPRINOQ-HWKANZROSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Ethyl trans-2-butenoate is a volatile flavor compound found in fruits such as cashew apple, African star apple, naranjilla fruit and sweet passion fruit.

Application


  • Comparison of the Fruit Volatile Profiles of Five Muscadine Grape Cultivars (Vitis rotundifolia Michx.) Using HS-SPME-GC/MS Combined With Multivariate Statistical Analysis.: This study details the analysis of volatile profiles in different Muscadine grape cultivars, providing insight into their chemical compositions during various stages of fruit development, which could be crucial for enhancing flavor and aroma in food products (Deng et al., 2021).

  • Effects of methanogenic inhibitors on methane production and abundances of methanogens and cellulolytic bacteria in in vitro ruminal cultures.: This study explores the reduction of methane emissions through the use of inhibitors, contributing to more sustainable agricultural practices and environmental conservation (Zhou et al., 2011).

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Volatiles from naranjilla fruit (Solanum quitoense Lam.). GC/MS analysis and sensory evaluation using sniffing GC
Brunke EJ, et al.
Journal of Agricultural and Food Chemistry, 37(3), 746-748 (1989)
Evaluation of volatile flavour compounds from cashew apple (Anacardium occidentale L) juice by the Osme gas chromatography/olfactometry technique.
Garruti DS, et al.
Journal of the Science of Food and Agriculture, 83(14), 1455-1462 (2003)
Volatile Composition of Sweet Passion Fruit (Passiflora alata Curtis).
Mamede AM, et al.
Journal of Chemistry (2017)
Headspace solid-phase microextraction gas chromatography?mass spectrometry determination of volatile compounds in different varieties of African star apple fruit (Chrysophillum albidum).
Lasekan O, et al.
Food Chemistry, 141(3), 2089-2097 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service