Skip to Content
Merck
All Photos(1)

Key Documents

THS05131

ThalesNano CatCart® catalyst cartridge system, 70 mm L

Pd EnCat® 30

Synonym(s):

Heterogeneous catalysis system, safe Heterogeneous catalysis

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103817
NACRES:
NB.51

packaging

pkg of 1 ea

manufacturer/tradename

ThalesNano THS 05131

diam.

4 mm

Looking for similar products? Visit Product Comparison Guide

General description

CatCarts are suitable for use only with Thalesnano H-Cube reactors.

Legal Information

CatCart is a registered trademark of ThalesNano Nanotechnology Inc.
EnCat is a registered trademark of Reaxa Ltd.

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Flam. Sol. 1 - Repr. 1B - Skin Irrit. 2 - STOT RE 2 Inhalation

Target Organs

Nervous system

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service