Key Documents
SYND251019
Synthware Vigreux microscale distillation head
joint: ST/NS 19/22, immersion depth 25 mm
Synonym(s):
Synthware D251019, short path, distilling head
About This Item
UNSPSC Code:
41104812
NACRES:
NB.51
Pricing and availability is not currently available.
Joint
joint: ST/NS19/22
packaging
pkg of 1 ea
manufacturer/tradename
Synthware D251019
immersion depth
25 mm
Related Categories
General description
With Vigreux Indentations, used with 25 mm immersion thermometer, and D27 distillation receiver.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)