SLW718242
Pyrex® Mortar
120 mL
Synonym(s):
Pyrex, SciLabware 718242
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.35
Recommended Products
manufacturer/tradename
718242
SciLabware
H
75 mm
O.D.
100 mm
capacity
120 mL
Looking for similar products? Visit Product Comparison Guide
General description
- Manufactured from Pyrex borosilicate glass for highest resistance to thermal shock and chemical corrosion
- For use with Pyrex Pestle
- Ideal for preparing compounds, mixing and grinding powders
Legal Information
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service