Skip to Content
Merck
All Photos(1)

Key Documents

S0251

Sigma-Aldrich

Sodium metaborate tetrahydrate

≥99%

Synonym(s):

Boric acid, sodium salt, tetrahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaBO2 · 4H2O
CAS Number:
Molecular Weight:
137.86
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
form:
powder
Pricing and availability is not currently available.

Quality Level

Assay

≥99%

form

powder

SMILES string

O.O.O.O.[Na+].[O-]B=O

InChI

1S/BO2.Na.4H2O/c2-1-3;;;;;/h;;4*1H2/q-1;+1;;;;

InChI key

JAKYJVJWXKRTSJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sodium metaborate tetrahydrate is a colorless crystalline solid that serves as a Boron source in the synthesis of NaBH4. It finds applications in the textile industry, as well as in flame retardants and additives.

Application

Sodium metaborate tetrahydrate can be used:

  • As a precursor to prepare an environment-friendly NaBO2/PMS catalytic system for efficient removal of organic pollutants.
  • To enhance the flame-retardant properties of epoxy fireproof coatings.

Caution

Readily reacts with CO2 in air.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alissa M Weaver et al.
Methods in molecular biology (Clifton, N.J.), 1046, 171-189 (2013-07-23)
Invasion by cancer cells through the extracellular matrix (ECM) of tissues is a critical step in cancer progression and metastasis. Actin-rich subcellular protrusions known as invadopodia are thought to facilitate this process by localizing proteinases which degrade the ECM and
Raquel Morais de Paiva Daibert et al.
PloS one, 15(11), e0241861-e0241861 (2020-11-07)
Macrophages are classified upon activation as classical activated M1 and M2 anti-inflammatory regulatory populations. This macrophage polarization is well characterized in humans and mice, but M1/M2 profile in cattle has been far less explored. Bos primigenius taurus (taurine) and Bos

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service