Skip to Content
Merck
All Photos(2)

Key Documents

H16205

Sigma-Aldrich

Homophthalic acid

98%

Synonym(s):

α-Carboxy-o-toluic acid, 2-Carboxyphenylacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO2CCH2C6H4CO2H
CAS Number:
Molecular Weight:
180.16
Beilstein:
1872069
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

mp

178-182 °C (lit.)

SMILES string

OC(=O)Cc1ccccc1C(O)=O

InChI

1S/C9H8O4/c10-8(11)5-6-3-1-2-4-7(6)9(12)13/h1-4H,5H2,(H,10,11)(H,12,13)

InChI key

ZHQLTKAVLJKSKR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Remainder phthalic acid

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T B Karegoudar et al.
FEMS microbiology letters, 49(2-3), 305-308 (1989-04-01)
A microorganism capable of degrading homophthalic acid as a sole source of carbon was isolated from soil. The strain was tentatively identified as Pseudomonas sp. Oxygen uptake studies were carried out with possible intermediates. Assays for several different enzymes were
Khalid Mohammed Khan et al.
Natural product research, 22(13), 1120-1127 (2008-10-16)
A microwave-assisted, environmentally friendly, high-yielding, time-saving synthesis of medicinally important 3-substituted isocoumarins was carried out in a single step by direct condensation of homophthalic acid with aryol and acyl chlorides under solvent-free conditions without any solid support. The synthesised isocoumarins
C S Karigar et al.
FEMS microbiology letters, 110(1), 59-64 (1993-06-01)
A microorganism capable of degrading homophthalic acid as a sole carbon source was isolated from garden soil. The strain was identified as Pseudomonas alcaligenes. The organism degraded homophthalate by a pathway which involved phenylacetate and p-hydroxyphenylacetate as intermediates. The intermediates

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service