Skip to Content
Merck
All Photos(2)

Key Documents

GF15342016

Platinum

tube, 500mm, outside diameter 1.2mm, inside diameter 1.0mm, wall thickness 0.1mm, as drawn, 99.95%

Synonym(s):

Platinum, PT007220

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Pt
CAS Number:
Molecular Weight:
195.08
MDL number:
UNSPSC Code:
12141734
PubChem Substance ID:
NACRES:
NA.23

Assay

99.95%

form

tubes

manufacturer/tradename

Goodfellow 153-420-16

resistivity

10.6 μΩ-cm, 20°C

L × thickness

500 mm × 0.1 mm

O.D. × I.D.

1.2 mm × 1.0 mm

bp

3827 °C (lit.)

mp

1772 °C (lit.)

density

21.45 g/cm3 (lit.)

SMILES string

[Pt]

InChI

1S/Pt

InChI key

BASFCYQUMIYNBI-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiao-yong Shen et al.
PloS one, 8(8), e69553-e69553 (2013-08-14)
X-ray repair cross-complementing protein 3 (XRCC3) is an essential gene involved in the double-strand break repair pathway. Published evidence has shown controversial results about the relationship between XRCC3 Thr241Met polymorphism and clinical outcomes of non-small cell lung cancer (NSCLC) patients
Josep Ramon Germà-Lluch et al.
Archivos espanoles de urologia, 66(5), 524-535 (2013-06-26)
Cisplatin has been the cornerstone of germ cell testicular tumors therapy since its introduction more tan 30 years ago, and a basic part of the schemes given to multiple ovarian, lung, head and neck, and bladder tumors among others. Some
Platinum-free catalysts as counter electrodes in dye-sensitized solar cells.
Mingxing Wu et al.
ChemSusChem, 5(8), 1343-1357 (2012-08-14)
Yanlong Yang et al.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine, 35(4), 2905-2921 (2013-12-18)
The relationship between the ERCC1/2 single nucleotide polymorphisms (SNPs) and the clinical outcomes of the platinum-based chemotherapy in the non-small cell lung cancer (NSCLC) is still inconsistent and inconclusive despite extensive investigations have been conducted to address this question. In
Xiaoyong Wang et al.
Chemical Society reviews, 42(1), 202-224 (2012-10-09)
Platinum-based anticancer drugs occupy a crucial role in the treatment of various malignant tumours. However, the efficacy and applicability of platinum drugs are heavily restricted by severe systemic toxicities and drug resistance. Different drug targeting and delivery (DTD) strategies have

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service