Skip to Content
Merck
All Photos(2)

Key Documents

GF04642332

Zinc

powder, max. particle size 150 micron, weight 100 g, high purity 99.999%

Synonym(s):

ZN006030

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Zn
CAS Number:
Molecular Weight:
65.39
MDL number:
UNSPSC Code:
12141750
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

1 mmHg ( 487 °C)

Assay

99.999%

form

powder

manufacturer/tradename

Goodfellow 046-423-32

resistivity

5.8 μΩ-cm, 20°C

weight

100 g

max. part. size

150 μm

bp

907 °C (lit.)

mp

420 °C (lit.)

density

7.133 g/mL at 25 °C (lit.)

SMILES string

[Zn]

InChI

1S/Zn

InChI key

HCHKCACWOHOZIP-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Pictograms

FlameEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Pyr. Sol. 1 - Self-heat. 1 - Water-react 1

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 6

1 of 6

Zinc powder, max. particle size 150 micron, weight 20 g, high purity 99.999%

GF09421095

Zinc

Zinc powder, mean particle size 7.5 micron, weight 100 g, purity 98.8%

GF20413718

Zinc

Zinc powder, max. particle size 150 micron, weight 100 g, purity 99.9%

GF52825743

Zinc

Zinc powder, 7.5 mean particle size (micron), weight 500 g, purity 98.8%

GF64346474

Zinc

Zinc powder, 5 max. part. size (micron), weight 100 g, min. particle size 1 micron

GF67222111

Zinc

Zinc powder, 150 max. part. size (micron), weight 1000 g, purity 99.9%

GF78560781

Zinc

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service