DWKBGF136P
Azlon Wash Bottles with Integral Spout
low-density polyethylene bottle, capacity 500 mL
Synonym(s):
BGF136P
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
13111000
NACRES:
NB.43
Recommended Products
material
low-density polyethylene bottle
packaging
pkg of 5 ea
manufacturer/tradename
Azlon BGF136P
capacity
500 mL
Looking for similar products? Visit Product Comparison Guide
General description
Featuring an ergonomic design, this AZLON® wash bottle with integral spout has an integrally moulded dispensing tube which minimises contamination events that occur during filling.
Application
Ideal for dispensing liquids
Features and Benefits
Rectangular design specifically suited for smaller hands
Legal Information
AZLON is a registered trademark of DWK Life Sciences
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service