Key Documents
CAD6512
1-way tubing connector
MLL to hose end for 1/4 in. to 5/16 in. I.D. tubing (316SS)
Synonym(s):
Tubing Connector
About This Item
UNSPSC Code:
40183110
NACRES:
NB.03
Pricing and availability is not currently available.
packaging
pack of 5 ea
manufacturer/tradename
Cadence Science 6512
General description
  • Plated brass or 316 stainless steel
  • Meets ISO 594 specifications
  • Final assembly water tested at 45 PSI
Female Luer Lock (FLL) - contains a tabbed hub for secure fitting to a threaded male Luer lock.
Micro-Mate® Female Luer Lock (MMFLL) - designed and manufactured to reduce the stripping of threads which occurs when metal adapters are mated with plastic male Luer lock adapters. Uniquely designed to facilitate easy cleaning and maintenance.
Male Luer Lock (MLL) - male Luer slip with a threaded sheath for a secure fitting to a female Luer lock.
Male Luer (ML) - male Luer slip that fits with a female Luer.
Legal Information
Micro-Mate is a registered trademark of Cadence Science
Choose from one of the most recent versions:
Certificates of Analysis (COA)