Skip to Content
Merck
All Photos(1)

Key Documents

C92006

Sigma-Aldrich

Indole-5-carbonitrile

99%

Synonym(s):

5-Cyanoindole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H6N2
CAS Number:
Molecular Weight:
142.16
Beilstein:
116738
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

powder

mp

106-108 °C (lit.)

SMILES string

N#Cc1ccc2[nH]ccc2c1

InChI

1S/C9H6N2/c10-6-7-1-2-9-8(5-7)3-4-11-9/h1-5,11H

InChI key

YHYLDEVWYOFIJK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Reactant for parallel synthesis of dihydroisoquinolines via silver and L-proline co-catalyzed three-component coupling reaction
  • Reactant for chemoselective and regioselective preparation of benzoyl indoles
  • Reactant for preparation of novel PPARα/γ dual agonists for potential treatment of metabolic syndrome and IDDM
  • Reactant for preparation of 4,5-dihydrocyclopenta[c]quinolines by palladium-catalyzed ring-expansion reaction alkynes, using O2 as the oxidant
  • Reactant for preparation of vinylindoles by hydroarylation of alkynes using indium bromide catalyst
  • Reactant for gold catalyzed direct alkynylation using a benziodoxolone based hypervalent iodine reagent

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Josefin Wilke et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 17(17), 2736-2743 (2016-06-02)
The estimate of the magnitude and the orientation of molecular electric dipole moments from the vector sum of bond or fragment dipole moments is a widely used approach in chemistry. However, the limitations of this intuitive model have rarely been

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service