Skip to Content
Merck
All Photos(1)

Key Documents

BR704526

BRAND® 8-channel manifold

polypropylene, for QuikSip, BT-Aspirator

Synonym(s):

Accessories, Aspirator, Bottle-top Dispenser, Dispenser, QuikSip

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121500
NACRES:
NB.01
Pricing and availability is not currently available.

material

polypropylene

feature

autoclavable

packaging

pack of 1 ea

manufacturer/tradename

BRAND 704526

Related Categories

General description

The QuikSip™ bottle-top aspirator from BRAND is designed for safe and fast aspiration of common laboratory liquids used in biology, food chemistry, and medicine. Use this 8-channel manifold for aspirating out of multiwell plates.

Application

Aspiration

Features and Benefits

Overview:
  • Safe removal of supernatants (up to max. 25 ml per plunger stroke) e.g. biological solutions, nutrient media, polar solvents, aqueous soutions.
  • Works without vacuum pump.
  • Fingertip vacuum control using the cell-culture unit.
  • For use with disposable pipette tips, micropipettes and glass pasteur pipettes.
  • Adapter and suction tube of the cell-culture are autoclavable at 121 °C (2 bar), acc. DIN EN 285, dispensing cartridge and pump unit are not autoclavable.


Limitations of use:
  • Vapor pressure up to 500 mbar
  • Density up to 2.2 g/cm³
  • Use between +15°C and +40°C
  • Viscosity 500 mm² /s (dynamic viscosity [mPas] = kinematic viscosity [mm² /s] x density [g/cm³)

Legal Information

BRAND is a registered trademark of BRAND GMBH + CO KG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service