ALD00592
N-Boc-N′-TFA-pyrazole-1-carboxamidine
95%
Synonym(s):
N-Boc,N′-trifluoroacetyl-guanylpyrazole, tert-Butyl-((1H-pyrazol-1-yl)(2,2,2-trifluoroacetamido)methylene)carbamate, TurboGuan
About This Item
Recommended Products
Quality Level
Assay
95%
form
powder or crystals
mp
120-125 °C
application(s)
peptide synthesis
storage temp.
−20°C
Application
Other Notes
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Baran Group offers zinc-based reagents for versatile transformations in organic synthesis.
Baran Group offers zinc-based reagents for versatile transformations in organic synthesis.
Baran Group offers zinc-based reagents for versatile transformations in organic synthesis.
Baran Group offers zinc-based reagents for versatile transformations in organic synthesis.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service