Skip to Content
Merck
All Photos(2)

Key Documents

932094

Sigma-Aldrich

TCzTrz

≥99% (HPLC)

Synonym(s):

9,9′,9"-(5-(4,6-Diphenyl-1,3,5-triazin-2-yl)benzene-1,2,3-triyl) tris(9H-carbazole)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C57H36N6
CAS Number:
Molecular Weight:
804.94
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.23
grade:
sublimed grade
solubility:
toluene: soluble

grade

sublimed grade

Quality Level

description

Triplet Energy:-2.80 eV

S1: 2.96 eV

T1: 2.81 eV

Assay

≥99% (HPLC)

energy gap

0.16 eV, delta EST

solubility

toluene: soluble

Orbital energy

HOMO -5.40 eV 
LUMO -2.18 eV 

UV absorption

λ: 335 nm±5 nm Amax
λ: 375 nm±5 nm Amax

Application

CzTrz exhibits good film-forming properties, high charge carrier mobility, and excellent triplet energy level compatibility with a wide range of phosphorescent and fluorescent dopants. These characteristics contribute to efficient charge recombination and emission of high-quality light in organic light emitting diodes (OLEDs). It can act as a hole-transporting material in the active layer of OFET devices, enabling efficient charge transport and modulation of conductivity.
Dopant for Light Blue Thermally Activated Delayed Fluorescence (TADF) OLED with EQE of 25%.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dong Ryun Lee et al.
Advanced materials (Deerfield Beach, Fla.), 27(39), 5861-5867 (2015-08-27)
Carbazole- and triazine-derived thermally activated delayed fluorescent (TADF) emitters, with three donor units and an even distribution of the highest occupied molecular orbital, achieve high external quantum efficiencies of above 25% in blue and green TADF devices.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service